Learn More
MRPL10, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human MRPL10 peptide using ARM Technology.
Supplier: Abnova Corporation H00124995K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. A pseudogene corresponding to this gene is found on chromosome 5q. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
MRPL10 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
MRPL10 | |
MRPL10 | |
A synthetic peptide of human MRPL10 | |
RUO | |
124995 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
mitochondrial ribosomal protein L10 | |
MRPL10 | |
L10MT, MGC17973, MRP-L10, MRP-L8, MRPL8, RPML8 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.