Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mitochondrial ribosomal protein L21, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89008681 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-008-681 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-008-681 Supplier Abnova Corporation Supplier No. H00219927B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human MRPL21 protein.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. Multiple transcript variants encoding different isoforms were identified through sequence analysis although some may be subject to nonsense-mediated decay (NMD). [provided by RefSeq

Sequence: MAAAMAASSLTVTLGRLASACSHSILRPSGPGAASLWSASRRFNSQSTSYLPGYVPKTSLSSPPWPEVVLPDPVEETRHHAEVVKKVNEMIVTGQYGRLFAVVHFASRQWKVTSEDLILIGNELDLACGERIRLEKVLLVGADNFTLLGKPLLGKDLVRVEATVIEKTESWPRIIMRFRKRKNFKKKRIVTTPQTVLRINSIEIAPCLL

Specifications

Antigen mitochondrial ribosomal protein L21
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human MRPL21 protein.
Formulation No additive
Gene MRPL21
Gene Accession No. BC055088
Gene Alias L21mt/MGC62013/MRP-L21
Gene Symbols MRPL21
Host Species Mouse
Immunogen MRPL21 (AAH55088, 1 a.a. ∼ 209 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 219927
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.