Learn More
mitochondrial ribosomal protein S27, Mouse, Clone: 3G8, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant MRPS27.
Supplier: Abnova Corporation H00023107M14A
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that may be a functional partner of the death associated protein 3 (DAP3). [provided by RefSeq
Sequence: SEEQSQNDEDNQGSEKLVEQLDIEETEQSKLPQYLERFKALHSKLQALGKIESEGLLSLTTQLVKEKLSTCEAEDIATYEQNLQQWHLDLVQLIQREQQQREQAKQEYQAQKAAKASASpecifications
mitochondrial ribosomal protein S27 | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
BC030521 | |
MRPS27 | |
MRPS27 (AAH30521, 51 a.a. ∼ 168 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
Yes | |
23107 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Western Blot | |
3G8 | |
Mouse monoclonal antibody raised against a full length recombinant MRPS27. | |
MRPS27 | |
FLJ21764/FLJ23348/KIAA0264/MRP-S27/S27mt | |
Mouse | |
200 μL | |
Stem Cell Biology | |
Primary | |
Human | |
Antibody | |
IgM κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.