Learn More
mitochondrial ribosomal protein S35, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human MRPS35 protein.
Supplier: Abnova Corporation H00060488B01
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that has had confusing nomenclature in the literature. Pseudogenes corresponding to this gene are found on chromosomes 3p, 5q, and 10q. [provided by RefSeq
Sequence: MAKEGNLELLKIPNFLHLTPVAIKKHCEALKDFCTEWPAALDSDEKCEKHFPIEIDCTDYVSSGPSVRNPRARVVVLRVKLSSLNLDDHAKKKLIKLVGERYCKTTDVLTIKTDRCPLRRQNYDYAVYLLTVLYHESWNTEEWEKSKTEADMEEYIWENSSSERNILETLLQMKAAEKNMEINKEELLGTKEIEEYKKSVVSLKNEEENENSISQYKESVKRLLNVTSpecifications
mitochondrial ribosomal protein S35 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human MRPS35 protein. | |
MRPS35 | |
DKFZp762P093/HDCMD11P/MDS023/MGC104278/MRP-S28/MRPS28 | |
Mouse | |
50 μL | |
Yes | |
60488 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
No additive | |
BC015862 | |
MRPS35 | |
MRPS35 (AAH15862, 1 a.a. ∼ 227 a.a) full-length human protein. | |
RUO | |
Primary | |
Human | |
Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.