Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRPS5, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017160 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-160 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-160 Supplier Abnova Corporation Supplier No. H00064969B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human MRPS5 protein.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that belongs to the ribosomal protein S5P family. Pseudogenes corresponding to this gene are found on chromosomes 4q, 5q, and 18q. [provided by RefSeq

Sequence: MATAVRAVGCLPVLCSGTAGHLLGRQCSLNTLPAASILAWKSVLGNGHLSSLGTRDTHPYASLSRALQTQCCISSPSHLMSQQYRPYSFFTKLTADELWKGALAETGAGAKKGRGKRTKKKKRKDLNRGQIIGEGRYGFLWPGLNVPLMKNGAVQTIAQRSKEEQEKVEADMIQQREEWDRKKKMKVKRERGWSGNSWGGISLGPPDPGPCGETYEDFDTRILEVRNVFTMTAKEGRKKSIRVLVAVGNGKGAAGFSIGKATDRMDAFRKAKNRAVHHLHYIERYEDHTIFHDISLRFKRTHIKMKKQPKGYGLRCHRAIITICRLIGIKDMYAKVSGSINMLSLTQGLFRGLSRQETHQQLADKKGLHVVEIREECGPLPIVVASPRGPLRKDPEPEDEVPDVKLDWEDVKTAQGMKRSVWSNLKRAAT

Specifications

Antigen MRPS5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human MRPS5 protein.
Formulation PBS with no preservative; pH 7.4
Gene MRPS5
Gene Accession No. NM_031902.3
Gene Alias MRP-S5/S5mt
Gene Symbols MRPS5
Host Species Mouse
Immunogen MRPS5 (NP_114108.1, 1 a.a. ∼ 430 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64969
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.