Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-MSH2 Clone: GB12,

Catalog No. NA26100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
NA26100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NA26100UG Supplier MilliporeSigma™ Supplier No. NA26100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects MSH2 Clone: GB12 in Human samples, and it is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Immuno Blotting

The MSH2 gene is one of at least 4 genes encoding proteins involved in the repair of mismatch nucleotides following DNA replication or repair. Mutations in the MSH2 gene contribute to the development of sporadic colorectal carcinoma, while germline MSH2 mutations are responsible for ∽50% of inherited, non-polyposis colorectal carcinoma (HNPCC). MSH2 encodes a protein of 934 amino acids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSH2
Applications Immunoblot, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone GB12
Conjugate Unconjugated
Formulation In 50mM sodium phosphate buffer, 0.2% gelatin, pH 7.5.
Host Species Mouse
Immunogen an N-terminal fragment of human MSH2
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽100kDa MSH2 protein in HCT116 and SW480 cells and colon tissue.
Target Species Human
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.