Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSH3 Mouse anti-Mouse, Clone: 1F6, MilliporeSigma™

Catalog No. MABE324MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE324MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE324MI Supplier MilliporeSigma Supplier No. MABE324
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-MSH3 Antibody, clone 1F6 is validated for use in Western Blotting and Immunocytochemistry for the detection of MSH3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSH3
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 1F6
Concentration 1mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_034959
Gene Symbols Msh3, Rep-3
Host Species Mouse
Immunogen Recombinant protein corresponding to mouse MSH3.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.