Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSH3 Mouse anti-Human, Unlabeled, Clone: 52, BD

Catalog No. BDB611390 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611390 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611390 Supplier BD Biosciences Supplier No. 611390
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Bacterial mismatch DNA repair involves the MutL, MutH, and MutS proteins, which forms a complex that mediates excision repair. Mutations in or deficiencies of any of these proteins results in a mutator phenotype that is characterized by genetic instability. Human homologs of MutS include MSH2, MSH3, and MSH6. MSH2 forms heterodimers with MSH6 (hMutSα) or MSH3 (hMutSβ) that specifically bind single-mispaired nucleotides and a subset of insertion-deletion mismatches. In addition, these heterodimers have intrinsic ATPase activity that is regulated by mismatch binding. ADP-bound heterodimers bind mismatched nucleotides, while ATP-bound heterodimers do not. The role of MSH3 in genetic stability in human cells in unclear. However, MSH3 and MSH6 share roles in the control of mutation rates. Both participate in repair of replication errors containing base-base mismatches or 1-4 extra bases. The MSH3 gene is located upstream of the dihydrofolate reductase (DHFR) gene and is expressed at low levels in a variety of human tissues. Thus, MSH3 is a component of an adenosine nucleotide-regulated molecular switch whose activity is essential for classical nucleotide mismatch repair.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen MSH3
Applications Western Blot
Classification Monoclonal
Clone 52
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human MSH3 aa. 136-349
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.