Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSH6 Mouse, Unlabeled, Clone: 44, BD

Catalog No. BDB610919 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610919 150 μg
BDB610918 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610919 Supplier BD Biosciences Supplier No. 610919
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DNA mismatch repair in bacteria is carried out by the MutL, MutH, and MutS proteins. Initial binding of MutS to the mismatched DNA is followed by binding of the MutH endonuclease and MutL. Together these proteins form a complex that mediates excision repair. Mutations or deficiencies of any of these bacterial proteins results in a mutator phenotype that is characterized by genetic instability. MSH2, MSH3 and MSH6 are human homologs of MutS, while MLH1, PMS1, and PMS2 are homologs of MutL. As a heterodimer with MSH2, MSH6 binds to DNA containing G/T mismatches. The MSH2-MSH6 complex recognizes single-base mispairs and insertion/deletion loops. Binding of this complex induces conformational changes in the DNA that lead to the binding of an MLH-PMS1 complex and excision repair. Mutations in the human genes are associated with hereditary nonpolyposis colon cancer (HNPCC), a common hereditary disease in humans. HNPCC is characterized by frequent microsatellite mutations that arise from somatic mutation due to a replication error (RER+) phenotype. This phenotype is analogous to the bacterial system and is directly linked to DNA mismatch repair deficiencies.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen MSH6
Applications Western Blot
Classification Monoclonal
Clone 44
Concentration 250μg/mL
Conjugate Unconjugated
Description GTBP
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human MSH6 aa. 225-333
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.