Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

musashi homolog 1 (Drosophila), Mouse, Clone: 3F2, Abnova™

Catalog No. 89000813 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-813 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-813 Supplier Abnova Corporation Supplier No. H00004440M04
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant MSI1.

This gene encodes a protein containing two conserved tandem RNA recognition motifs. Similar proteins in other species function as RNA-binding proteins and play central roles in posttranscriptional gene regulation. Expression of this gene has been correlated with the grade of the malignancy and proliferative activity in gliomas and melanomas. A pseudogene for this gene is located on chromosome 11q13. [provided by RefSeq

Sequence: METDAPQPGL ASPDSPHDPC KMFIGGLSWQ TTQEGLREYF GQFGE VKECL VMRDPLTKRS RGFGFVTFMD QAGVDKVLAQ SRHELDS

Specifications

Antigen musashi homolog 1 (Drosophila)
Applications ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 3F2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant MSI1.
Formulation PBS with no preservative; pH 7.4
Gene MSI1
Gene Accession No. NM_002442
Gene Symbols MSI1
Host Species Mouse
Immunogen MSI1 (NP_002433, 1 a.a. ∼ 87 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 4440
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.