Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSI1 Mouse anti-Human, PE, Clone: N14-47, BD

Catalog No. BDB561468 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561468 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561468 Supplier BD Biosciences Supplier No. 561468
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The N14-47 monoclonal antibody recognizes RNA-binding protein Musashi homolog 1 (Musashi-1 or MSI1) that represses the translation of particular mRNAs by binding specific RNA sequence motifs. MSI1 contains two RNA-recognition domains at amino acids 20-110 and 109-186. It is expressed in early lineage cells, such as neural stem/progenitor cells, small intestinal stem cells, and mammary stem cells. MSI1 has also been detected in many human cancers, including gliomas, melanomas, colorectal adenomas and adenocarcinomas, suggesting its involvement in cancer development.

TRUSTED_SUSTAINABILITY

Specifications

Antigen MSI1
Applications Flow Cytometry
Classification Monoclonal
Clone N14-47
Conjugate PE
Description Musashi-1, RNA-binding protein Musashi homolog 1, MSI1H
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human MSI1 a.a. 221-311 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.