Learn More
musashi homolog 2 (Drosophila), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human MSI2 protein.
Supplier: Abnova Corporation H00124540B01
Description
This gene encodes a protein containing two conserved tandem RNA recognition motifs. Similar proteins in other species function as RNA-binding proteins and play central roles in posttranscriptional gene regulation. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq
Sequence: MEANGSQGTSGSANDSQHDPGKMFIGGLSWQTSPDSLRDYFSKFGEIRECMVMRDPTTKRSRGFGFVTFADPASVDKVLGQPHHELDSKTIDPKVAFPRRAQPKMVTRTKKIFVGGLSANTVVEDVKQYFEQFGKVEDAMLMFDKTTNRHRGFGFVTFENEDVVEKVCEIHFHEINNKMVECKKAQPKEVMFPPGTRGRARGLPYTMDAFMLGMGMLGYPNFVATYGRGYPGFAPSYGYQFPGFPAAAYGPVAAAAVAAARGSGSNPARPGGFPGANSPGPVADLYGPASQDSGVGNYISAASPQPGSGFGHGIAGPLIATAFTNGYHSpecifications
musashi homolog 2 (Drosophila) | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human MSI2 protein. | |
MSI2 | |
FLJ36569/MGC3245/MSI2H | |
Mouse | |
50 μL | |
Yes | |
124540 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
No additive | |
NM_138962.2 | |
MSI2 | |
MSI2 (NP_620412.1, 1 a.a. ∼ 328 a.a) full-length human protein. | |
RUO | |
Primary | |
Human | |
Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.