Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSX2 Mouse anti-Human, Rat, Mouse, Clone: 7D11.1, MilliporeSigma™

Catalog No. MABD22MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABD22MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABD22MI Supplier MilliporeSigma Supplier No. MABD22

Mouse Monoclonal Antibody

Anti-MSX2 Antibody, clone 7D11.1 is an antibody against MSX2 for use in western blot, flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MSX2
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 7D11.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_002440
Gene Symbols MSX2, HOX8
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human MSX2.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.