Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MT1-MMP catalytic domain Mouse anti-Human, Clone: 3G4.2, MilliporeSigma™

Catalog No. MAB1767MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1767MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1767MI Supplier MilliporeSigma Supplier No. MAB1767
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Human MT1-MMP catalytic domain using this Anti-Human MT1-MMP catalytic domain Antibody, clone 3G4.2 validated for use in western blot, immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MT1-MMP catalytic domain
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 3G4.2
Conjugate Unconjugated
Formulation Protein A purified immunoglobulin liquid in PBS, pH 7.2, containing 0.1% sodium azide.
Gene Accession No. NM_004995.2
Gene Symbols MMP14, MTMMP1, MMP-14, MT1MMP, MT1-MMP, MMP-X1, EC 3.4.24.80
Host Species Mouse
Immunogen Recombinant protein containing the catalytic domain of human MT1-MMP.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.