Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-MT1-MMP Clone: 113-5B7,

Catalog No. IM57100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
IM57100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IM57100UG Supplier MilliporeSigma™ Supplier No. IM57100UG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Specifically detects MT1-MMP Clone: 113-5B7 in Human, Mouse, Rabbit, Rat samples, and it is validated for Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation

Matrix metalloproteinases (MMPs) are a family of enzymes that are responsible for the degradation of extracellular matrix components. Of the sixteen proteins reported to date, ten are normally found as soluble molecules. Several of the MMP proteins have been shown to be integral membrane proteins and have been named MT-MMPs for membrane bound MMP. The MT-MMP family is now known to contain at least three members, MT1-MMP, MT2-MMP and MT3-MMP, also known as MMP14, MMP15, and MMP16 respectively. While each of these proteins contain a C-terminal transmembrane domain allowing localization to the cell surface they are independent in expression. These proteins also differ from the other members of the MMP family in that they contain an 8 amino acid insert in the catalytic domain. The MT1-MMP protein is encoded by a 4.5 kb mRNA species giving rise to a protein with a molecular weight of 60-66kDa by SDS-PAGE. MT1-MMP is responsible for cleaving progelatinase A (MMP-2, 72kDa Type IV collagenase) and progelatinase B to their active forms. MT1-MMP itself requires an activation step that is the result of the activity of the membrane plasmin cascade. MT1-MMP functions by binding TIMP-2 and then the COOH terminal end of MMP-2 resulting in a 105kDa trimer which effects the cleavage of pro-MMP-2 to the biologically active form. The order of the binding of pro-MMP-2 and TIMP-2 to MT1-MMP is critical as TIMP-2 will also inhibit the activity of MMP-2 when present in a soluble form.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MT1-MMP
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone 113-5B7
Conjugate Unconjugated
Formulation In 100mM sodium phosphate buffer, 0.1% BSA, pH 7.0.
Host Species Mouse
Immunogen a synthetic peptide (CDGNFDTVAMLRGEM) corresponding to amino acids 319-333 in the hemopexin-like domain of human MT1-MMP
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽60kDa MTI-MMP protein in HT-1080 cells and in ConA-treated MD-MB-231 cells.
Target Species Human, Mouse, Rabbit, Rat
Content And Storage −20°C
Form Purified
Isotype IgG3
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.