Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MT1-MMP Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB6004MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB6004MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB6004MI Supplier MilliporeSigma Supplier No. AB6004
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect MT1-MMP using this Anti-MT1-MMP Antibody, hinge region validated for use in western blot, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MT1-MMP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified in 0.1M Tris-Glycine, pH 7.4, 150mM NaCl with 0.05% NaN3.
Gene Symbols EC 3.4.24.80 ; MMP-14 ; MMP-X1 ; MT1-MMP ; MT1MMP; MTMMP1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide selected from the hinge region of human MT1-MMP.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) P50281
Target Species Human, Rat
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.