Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-MTA3 (400-450), Polyclonal

Catalog No. IM101250UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
IM101250UG 50 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IM101250UG Supplier MilliporeSigma™ Supplier No. IM101250UG

Rabbit Polyclonal Antibody

Specifically detects MTA3 (400-450) Clone: in Human samples, and it is validated for Immunocytochemistry, Immunoprecipitation

MTA1, MTA2, and MTA3 (Metastasis-associated gene 3, ACC# AAH53631) are members of a growing novel gene family associated with highly metastatic human carcinomas. MTA3 is expressed in all tissues as both 515 aa (MTA3) and 594 aa (MTA3L) proteins found in both the nucleus and the cytoplasmic compartments. It was identified as an Estrogen receptor (ER) regulated component of the Mi/NuRD histone deacetylase transcriptional repressor. In response to estrogen, MTA3 directly targets Snail, the master regulator of the epithelial to mesenchymal transition (EMT). Regulation of E-cadherin expression due to MTA3 and ER mediated signaling of Snail expression may be an important pathway in human breast tumors. Loss of ER results in decreased MTA3 expression and tamoxifen reduces expression of MTA3 in endometrial cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MTA3 (400-450)
Applications Immunocytochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation In Tris-citrate/phosphate buffer, pH 7.0-8.0.
Host Species Rabbit
Immunogen a synthetic peptide corresponding to amino acids within residues 400-450 of human MTA3
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽62kDa MTA3 protein in MCF-7 cells.
Target Species Human
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.