Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
metastasis associated 1 family, member 3, Mouse, Clone: 3B3, Abnova™
Mouse monoclonal antibody raised against a partial recombinant MTA3.
Supplier: Abnova Corporation H00057504M02
Description
Sequence: LKMPTQSEEEKLSPSPTTEDPRVRSHVSRQAMQGMPVRNTGSPKSAVKTRQAFFLHTTYFTKFARQVCKNTLRLRQAARRPFVAINYAAIRAECKMLLNSSpecifications
metastasis associated 1 family, member 3 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_020744 | |
MTA3 | |
MTA3 (NP_065795, 416 a.a. ∼ 515 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Transcription Regulation | |
Primary | |
Human | |
Antibody |
ELISA, Immunofluorescence | |
3B3 | |
Mouse monoclonal antibody raised against a partial recombinant MTA3. | |
MTA3 | |
KIAA1266 | |
Mouse | |
Affinity Purified | |
RUO | |
Yes | |
57504 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction