Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

metal-regulatory transcription factor 1, Mouse, Clone: 2C12, Abnova™

Catalog No. 89000818 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-818 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-818 Supplier Abnova Corporation Supplier No. H00004520M04
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant MTF1.

This gene encodes a transcription factor that induces expression of metallothioneins and other genes involved in metal homeostasis in response to heavy metals such as cadmium, zinc, copper, and silver. The protein is a nucleocytoplasmic shuttling protein that accumulates in the nucleus upon heavy metal exposure and binds to promoters containing a metal-responsive element (MRE). [provided by RefSeq

Sequence: GTVYDRTTVLIEQDPGTLEDEDDDGQCGEHLPFLVGGEEGFHLIDHEAMSQGYVQHIISPDQIHLTINPGSTPMPRNIEGATLTLQSECPETKRKEVKRY

Specifications

Antigen metal-regulatory transcription factor 1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2C12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant MTF1.
Formulation PBS with no preservative; pH 7.4
Gene MTF1
Gene Accession No. NM_005955
Gene Alias MGC23036/MTF-1/ZRF
Gene Symbols MTF1
Host Species Mouse
Immunogen MTF1 (NP_005946, 41 a.a. ∼ 140 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 4520
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.