Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTHFD1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89002949 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-949 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-949 Supplier Abnova Corporation Supplier No. H00004522D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human MTHFD1 protein.

This gene encodes a protein that possesses three distinct enzymatic activities, 5,10-methylenetetrahydrofolate dehydrogenase, 5,10-methenyltetrahydrofolate cyclohydrolase and 10-formyltetrahydrofolate synthetase. Each of these activities catalyzes one of three sequential reactions in the interconversion of 1-carbon derivatives of tetrahydrofolate, which are substrates for methionine, thymidylate, and de novo purine syntheses. The trifunctional enzymatic activities are conferred by two major domains, an aminoterminal portion containing the dehydrogenase and cyclohydrolase activities and a larger synthetase domain. [provided by RefSeq

Sequence: MAPAEILNGKEISAQIRARLKNQVTQLKEQVPGFTPRLAILQVGNRDDSNLYINVKLKAAEEIGIKATHIKLPRTTTESEVMKYITSLNEDSTVHGFLVQLPLDSENSINTEEVINAIAPEKDVDGLTSINAGRLARGDLNDCFIPCTPKGCLELIKETGVPIAGRHAVVVGRSKIVGAPMHDLLLWNNATVTTCHSKTAHLDEEVNKGDILVVATGQPEMVKGEWIKPGAIVIDCGINYVPDDKKPNGRKVVGDVAYDEAKERASFITPVPGGVGPMTVAMLMQSTVESAKRFLEKFKPGKWMIQYNNLNLKTPVPSDIDISRSCKPKPIGKLAREIGLLSEEVELYGETKAKVLLSALERLKHRPDGKYVVVTGITPTPLGEGKSTTTIGLVQALGAHLYQNVFACVRQPSQGPTFGIKGGAAGGGYSQVIPMEEFNLHLTGDIHAITAANNLVAAAIDARIFHELTQTDKALFNRLVPSVNGVRRFSDIQIRRLKRLGIEKTDPTTLTDEEINRFARLDIDPETITWQRVLDTNDRFLRKITIGQAPTEKGHTRTAQFDISVASEIMAVLALTTSLEDMRERLGKMVVASSKKGEPVSAEDLGVSGALTVLMKDAIKPNLMQTLEGTPVFVHAGPFANIAHGNSSIIADRIALKLVGPEGFVVTEAGFGADIGMEKFFNIKCRYSGLCPHVVVLVATVRALKMHGGGPTVTAGLPLPKAYIQENLELVEKGFSNLKKQIENARMFGIPVVVAVNAFKTDTESELDFISRLSREHGAFDAVKCTHWAEGGKGALALAQAVQRAAQAPSSFQLLYDLKLPVEDKIRIIAQKIYGADDIELLPEAQHKAEVYTKQGFGNLPICMAKTHLSLSHNPEQKGVPTGFILPIRDIRASVGAGFLYPLVGTMSTMPGLPTRPCFYDIDLDPETEQVNGLF

Specifications

Antigen MTHFD1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human MTHFD1 protein.
Formulation PBS with no preservative; pH 7.4
Gene MTHFD1
Gene Accession No. BC050420.1
Gene Alias MTHFC/MTHFD
Gene Symbols MTHFD1
Host Species Rabbit
Immunogen MTHFD1 (AAH50420.1, 1 a.a. ∼ 935 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4522
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.