Learn More
MTMR1, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human MTMR1 peptide using ARM Technology.
Supplier: Abnova Corporation H00008776K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene encodes a member of the myotubularin related family of proteins. Members of this family contain the consensus sequence for the active site of protein tyrosine phosphatases. Alternatively spliced variants have been described but their biological validity has not been determined. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
MTMR1 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
MTMR1 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
1mg | |
myotubularin related protein 1 | |
MTMR1 | |
MTMR1 | |
A synthetic peptide of human MTMR1 | |
RUO | |
8776 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.