Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

myotubularin related protein 3, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016958 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-958 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-958 Supplier Abnova Corporation Supplier No. H00008897B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human MTMR3 protein.

This gene encodes a member of the myotubularin dual specificity protein phosphatase gene family. The encoded protein is structurally similar to myotubularin but in addition contains a FYVE domain and an N-terminal PH-GRAM domain. The protein can self-associate and also form heteromers with another myotubularin related protein. The protein binds to phosphoinositide lipids through the PH-GRAM domain, and can hydrolyze phosphatidylinositol(3)-phosphate and phosphatidylinositol(3,5)-biphosphate in vitro. The encoded protein has been observed to have a perinuclear, possibly membrane-bound, distribution in cells, but it has also been found free in the cytoplasm. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MDEETRHSLECIQANQIFPRKQLIREDENLQVPFLELHGESTEFVGRAEDAIIALSNYRLHIKFKESLVNVPLQLIESVECRDIFQLHLTCKDCKVIRCQFSTFEQCQEWLKRLNNAIRPPAKIEDLFSFAYHAWCMEVYASEKEQHGDLCRPGEHVTSRFKNEVERMGFDMNNAWRISNINEKYKLCGSYPQELIVPAWITDKELESVSSFRSWKRIPAVIYRHQSNGAVIARCGQPEVSWWGWRNADDEHLVQSVAKACASDSRSSGSKLSTRNTSRDFPNGGDLSDVEFDSSLSNASGAESLAIQPQKLLILDARSYAAAVANRAKGGGCECPEYYPNCEVVFMGMANIHSIRRSFQSLRLLCTQMPDPGNWLSALESTKWLHHLSVLLKSALLVVHAVDQDQRPVLVHCSDGWDRTPQIVALAKLLLDPYYRTIEGFQVLVEMEWLDFGHKFADRCGHGENSDDLNERCPVFLQWLDCVHQLQRQFPCSFEFNEAFLVKLVQHTYSCLFGTFLCNNAKERGEKHTQERTCSVWSLLRAGNKAFKNLLYSSQSEAVLYPVCHVRNLMLWSAVYLPCPSPTTPVDDSCAPYPAPGTSPDDPPLSRLPKTRSYDNLTTACDNTVPLASRRCSDPSLNEKWQEHRRSLELSSLAGPGEDPLSADSLGKPTRVPGGAELSVAAGVAEGQMENILQEATKEESGVEEPAHRAGIEIQEGKEDPLLEKESRRKTPEASAIGLHQDPELGDAALRSHLDMSWPLFSQGISEQQSGLSVLLSSLQVPPRGEDSLEVPVEQFRIEEIAEGREEAVLPIPVDAKVGYGTSQSCSLLPSQVPFETRGPNVDSSTDMLVEDKVKSVSGPQGHHRSCLVNSGKDRLPQTMEPSPSETSLVERPQVGSVVHRTSLGSTLSLTRSPCALPLAECKEGLVCNGAPETENRASEQPPGLSTLQMYPTPNGHCANGEAGRSKDSLSRQLSAMSCSSAHLHSRNLHHKWLHSHSGRPSATSSPDQPSRSHLDDDGMSVYTDTIQQRLRQIESGHQQEVETLKKQVQELKSRLESQYLTSSLHFNGDFGDEVTSIPDSESNLDQNCLSRCSTEIFSEASWEQVDKQDTEMTRWLPDHLAAHCYACDSAFWLASRKHHCRNCGNVFCSSCCNQKVPVPSQQLFEPSRVCKSCYSSLHPTSSSIDLELDKPIAATSN

Specifications

Antigen myotubularin related protein 3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human MTMR3 protein.
Formulation PBS with no preservative; pH 7.4
Gene MTMR3
Gene Accession No. NM_021090.2
Gene Alias FLJ32333/FYVE-DSP1/KIAA0371/ZFYVE10
Gene Symbols MTMR3
Host Species Mouse
Immunogen MTMR3 (NP_066576.1, 1 a.a. ∼ 1198 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8897
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.