Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-mTOR (catalystic domain), Polyclonal,

Catalog No. 071415MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-141-5MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-141-5MI Supplier MilliporeSigma™ Supplier No. 071415

Specifications

Antigen mTOR (catalystic domain)
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal IgG in PBS containing 10% glycerol with 0.02% sodium azide added as a preservative.
Gene Symbols FRAP1; RAPT1; FLJ44809; MTOR; FRAP; mTOR; FRAP2; RAFT1
Host Species Rabbit
Immunogen Recombinant GST fusion protein corresponding to amino acids 1223-1290 of human mTOR.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_004958.2
Target Species Human, Rat
Content And Storage −70°C for up to 12 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.