Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mTOR Mouse anti-Human, Rat, Clone: F11, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABS14
Description
This Anti-mTOR Antibody, clone F11 is validated for use in western blot, immunoprecipitation for the detection of mTOR.
Specifications
mTOR | |
Monoclonal | |
Unconjugated | |
NP_063971 | |
Mouse | |
Protein G | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Immunoprecipitation, Western Blot | |
F11 | |
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
FRAP, FRAP1, FRAP2, RAFT1, RAPT1, mTOR | |
GST-tagged recombinant protein corresponding to rat mTOR. | |
100 μg | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction