Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUC1/episialin Mouse anti-Human, Clone: 214D4, MilliporeSigma™

Catalog No. 05652MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-652-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-652-MI Supplier MilliporeSigma Supplier No. 05652
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-MUC1/episialin Antibody, clone 214D4 detects level of MUC1/episialin and has been published and validated for use in flow cytometry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MUC1/episialin
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 214D4
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Gene Accession No. NM_182741
Gene Symbols MUC1, MUC1/ZD, MUC-1/X, PEMT, MUC-1, Episialin, episialin, MUC-1/SEC, PUM, EMA, PEM, H23AG, MAM6, CD227
Host Species Mouse
Immunogen Fusion protein, human b-gal-episialin/MUC1 repeat domain
Purification Method Protein G chromatography
Quantity 200 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.