Learn More
MUC2 Mouse anti-Human, Unlabeled, Clone: CCP58, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 555926
Description
Reacts with the core peptide of the MUC2 protein, a member of a family of mucin glycoproteins that are characterized by high carbohydrate content, O-linked oligosaccharides, high molecular weight (>200kDa) and an amino acid composition rich in serine, threonin, proline and glycine. The core protein contains a domain of 23 amino-acid tandem repeats which function as multiple epitopes for the monoclonal antibody. Incomplete glycosylation of some tumor-associated mucins may lead to variable unmasking of the multiple peptide epitopes leading to the observed differences in staining intensity between normal and malignant tissues. This antibody has been shown to be relatively tissue specific, being associated predominantly with intestinal tissue and is highly reactive with colon and gastric tumors. The antibody is suitable for staining formalin-fixed, paraffin-embedded tissue sections following citrate pretreatment.
Host Species: Mouse
Clone: CCP58
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)

Specifications
MUC2 | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Immunohistochemistry | |
CCP58 | |
Unconjugated | |
Mouse | |
0.1 mg | |
Cell Biology | |
Human | |
IgG1 κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.