Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mucin 5B Mouse anti-Human, Monkey, Clone: 15.5B, MilliporeSigma™

Catalog No. MAB3828MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3828MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3828MI Supplier MilliporeSigma Supplier No. MAB3828
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Mucin 5B Antibody, clone 15.5B is validated for use in ELISA, western blot, immunohistochemistry for the detection of Mucin 5B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mucin 5B
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 15.5B
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in PBS containing 0.1% sodium azide.
Gene Accession No. NM_002458.1
Gene Symbols MUC5B, MG1, MUC5, MUC9
Host Species Mouse
Immunogen Un-glycosylated, serine- and threonine-free synthetic peptide from the central region of human MUC5B.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Monkey
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.