Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mucin MUC5AC Mouse anti-Human, Clone: CLH2, MilliporeSigma™

Catalog No. MAB2011MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2011MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2011MI Supplier MilliporeSigma Supplier No. MAB2011
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Mucin MUC5AC Antibody, clone CLH2 detects level of Mucin MUC5AC and has been published and validated for use in ELISA, immunoprecipitation, western blot, immunocytochemistry, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mucin MUC5AC
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone CLH2
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid at 1mg/mL in 0.02M phosphate buffer, 0.25Msodium chloride, 0.1% sodium azide, pH 7.6.
Gene Accession No. NM_005961.2
Gene Symbols MUC6
Host Species Mouse
Immunogen Directed against a synthetic peptide of the MUC5AC tandem repeat.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.