Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Multi-Drug Resistance Related Protein Mouse anti-Human, Clone: MRPm6, MilliporeSigma™

Catalog No. MAB4122MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4122MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4122MI Supplier MilliporeSigma Supplier No. MAB4122
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Multi-Drug Resistance Related Protein Antibody, clone MRPm6 is an antibody against Multi-Drug Resistance Related Protein for use in flow cytometry, western blot, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Multi-Drug Resistance Related Protein
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone MRPm6
Conjugate Unconjugated
Formulation Tissue Culture Supernatant. Liquid containing 0.1% sodium azide and 0.7% BSA.
Gene Accession No. NM_019901.1; NM_019902.1; NM_004996.3; NM_019862.2; NM_019898.2; NM_019899.2; NM_019900.2
Gene Symbols ABCC1, 4transporter), ABC29, GS-X, ABCC, MRP1, DKFZp781G125, MRP, DKFZp686N04233
Host Species Mouse
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots. Avoid repeated freeze/thaw cycles.
Form Supernatant
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.