Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mumps Blend Mouse anti-Human, Clone: 570-7A, 570-12G, MilliporeSigma™

Catalog No. MAB846MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB846MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB846MI Supplier MilliporeSigma Supplier No. MAB846
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Mumps Blend Antibody, fusion protein, clones 570-7A, 570-12G is an antibody against Mumps Blend for use in ELISA, immunofluorescence and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mumps Blend
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 570-7A, 570-12G, 570-12G
Conjugate Unconjugated
Formulation In 0.02M phosphate buffer, 0.25 NaCl with 0.1% sodium azide.
Host Species Mouse
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.