Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Munc-18 Mouse, Unlabeled, Clone: 31, BD

Catalog No. BDB610337 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610337 150 μg
BDB610336 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610337 Supplier BD Biosciences Supplier No. 610337
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

There are two Munc-18 isoforms: Munc-18-1 and Munc-18-2. Munc-18-1 (n-sec1 or rb-sec1; 68 kDa) is a neuronal protein that binds tightly to syntaxin 1 and functions in synaptic vesicle exocytosis. Munc-18-2 exhibits 63% amino acid sequence identity with Munc-18-1. Although Munc-18-2 is expressed in most tissues, Munc-18-1 is primarily expressed in brain. Both forms of Munc-18 bind tightly to syntaxins 1A, 2, and 3, but not to syntaxin 4. Syntaxin 1A is a plasma membrane protein implicated in synaptic vesicle docking. Following calcium entry into the presynaptic nerve terminal, the neurotransmitter-containing synaptic vesicle fuses with the plasma membrane at a region known as the active zone. Thus, by virtue of its interaction with syntaxin 1A, Munc-18-1 is thought to have an essential function in neurotransmitter release.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Munc-18
Applications Western Blot
Classification Monoclonal
Clone 31
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.