Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUPP1 Mouse, Unlabeled, Clone: 43, BD

Catalog No. BDB611558 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611558 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611558 Supplier BD Biosciences Supplier No. 611558

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

The postsynaptic density protein PSD-95 interacts with the cytoplasmic tail of ion channel subunits via a protein-protein motif called the PDZ domain. PSD-95 belongs to a family of PDZ domain-containing proteins that includes SAP97/Dlg, PSD-93/chapsyn-110, and SAP102. These proteins are characterized by three PDZ domains in the N-terminal half and an SH3 and guanylate kinase-like domain in the C-terminal region. They may be involved in the localization and clustering of ion channels and receptors at synaptic sites. Other PDZ domain-containing proteins such as glutamate receptor interacting protein (GRIP), Homer, and multi-PDZ-domain protein (MUPP1) do not contain guanylate kinase-like domains, but have also been implicated in receptor localization. MUPP1 contains thirteen PDZ domains and is expressed in human heart, brain, placenta, liver, skeletal muscle, kidney, and pancreas. In addition, smaller variants of MUPP1 are expressed in heart, liver, kidney, and brain. The C-terminal region of the 5-HT2c receptor interacts with MUPP1, suggesting that MUPP1 may have important protein-protein interactions during G-protein coupled receptor signaling.

Host Species: Mouse
Clone: 43
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Rat MUPP1 aa. 65-247
Formula Weight [Chemical]: 219kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen MUPP1
Applications Western Blot
Classification Monoclonal
Clone 43
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat MUPP1 aa. 65-247
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.