Learn More
Muscarinic Acetylcholine Receptor m1 Rabbit anti-Mouse, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Muscarinic Acetylcholine Receptor m1 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA and 0.05% sodium azide as a preservative. Reconstitute with 200μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 x |
| Gene Symbols | CHRM1; M1; HM1; MGC30125 |
| Host Species | Rabbit |
| Immunogen | GST fusion protein and part of i3 intercellular loop of human m1 muscarinic acetylcholine receptor (amino acids 227-353) (Accession P11229). |
| Purification Method | Affinity Purified |
| Quantity | 200 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.