Learn More
MilliporeSigma™ anti-Muscarinic Acetylcholine Receptor m2 ; Polyclonal; ™ (Chemicon™)
Supplier: MilliporeSigma™ AB5166200UL/DEL
Description
Rybin, 2000
Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
| Muscarinic Acetylcholine Receptor M2 | |
| Unconjugated | |
| GST fusion protein and part of i3 intercellular loop of human m2 musDrinic acetylcholine receptor (amino acids 225-356) | |
| 200 μL | |
| Neuroscience | |
| Human, Mouse, Rat |
| Polyclonal | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary |