Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Muscarinic Acetylcholine Receptor m4 Mouse anti-Monkey, Clone: 18C7.2, MilliporeSigma™

Catalog No. MAB1578MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1578MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1578MI Supplier MilliporeSigma Supplier No. MAB1578
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Muscarinic Acetylcholine Receptor m4 Antibody, clone 18C7.2 detects level of Muscarinic Acetylcholine Receptor m4 and has been published and validated for use in immunohistochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Muscarinic Acetylcholine Receptor m4
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 18C7.2
Conjugate Unconjugated
Formulation Liquid in 0.02M Phosphate buffer, 0.25MNaCl with 0.1% sodium azide.
Gene Accession No. NM_000741.2
Gene Symbols CHRM4, HM4
Host Species Mouse
Immunogen m4 receptor i3 loop (human), fused to GST
Quantity 100 μL
Primary or Secondary Primary
Target Species Monkey
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.