Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Muscle Phosphofructokinase/PFKM Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7687
This item is not returnable.
View return policy
Description
Muscle Phosphofructokinase/PFKM/PFK-1 Monoclonal specifically detects Muscle Phosphofructokinase/PFKM/PFK-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.Specifications
Muscle Phosphofructokinase/PFKM/PFK-1 | |
Monoclonal | |
LYOPH | |
Western Blot 0.2 ug/mL, Simple Western 2 ug/mL | |
P08237 | |
PFKM | |
E. coli-derived recombinant human Muscle Phosphofructokinase/PFKM Asn674-Val780 Accession # P08237 | |
100 μg | |
Primary | |
Detects human Muscle Phosphofructokinase/PFKM in direct ELISAs and Western blots. Detects mouse and rat Muscle Phosphofructokinase/PFKM in Western blots. | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
842735 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
6-phosphofructokinase, muscle type, EC 2.7.1,6-phosphofructo-1-kinase, EC 2.7.1.11, GSD7, MGC8699, PFK1, PFK-1, PFKA, PFK-A, PFKM, PFKX, Phosphofructo-1-kinase isozyme A, Phosphofructokinase 1, phosphofructokinase, muscle, phosphofructokinase, polypeptide X, Phosphofructokinase-1, Phosphofructokinase-M, phosphohexokinase | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5213 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction