Learn More
MBNL1, Invitrogen™

Mouse Monoclonal Antibody
Supplier: Heidolph North America 015908402
Description
Several important human diseases are associated with expansion of polynucleotide sequences, in most cases trinucleotide repeats. Myotonic dystrophy (DM1) is one of these diseases, and is associated with increases in the number of CTG repeats in the 3' UTR of the gene encoding myotonin (a.k.a. DM Kinase, DMK or myotonin-Protein Kinase), a ser/thr kinase expressed specifically in muscle. Mammalian genomes contain three muscleblind-like proteins, generally referred to as MBNL1, MBNL2 and MBNL3, and all three were found to associate with long double stranded CUG RNA repeats, thus, being triplet repeat expansion dsRNA-binding proteins.
Specifications
| MBNL1 | |
| Monoclonal | |
| Assay dependent | |
| Q9NR56, Q9JKP5 | |
| MBNL1 | |
| Recombinant human Muscleblind protein | |
| RUO | |
| 282635, 4154, 56758 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| MBNL1 | |
| Muscleblind-like protein 1, MBNL | |
| Mouse | |
| 500 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.