Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MUSK, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89205655 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-205-655 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-205-655 Supplier Abnova Corporation Supplier No. H00004593K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human MUSK peptide using ARM Technology.

Intercellular communication is often mediated by receptors on the surface of one cell that recognize and are activated by specific protein ligands released by other cells. Members of one class of cell surface receptors, receptor tyrosine kinases (RTKs), are characterized by having a cytoplasmic domain containing intrinsic tyrosine kinase activity. This kinase activity is regulated by the binding of a cognate ligand to the extracellular portion of the receptor. DeChiara et al. (1996) [PubMed 8653786] noted that the RTKs, known to be expressed in cell type-specific fashions, play a role critical for the growth and differentiation of those cell types. For example, members of the neural-specific TRK family that recognize nerve growth factor are absolutely required for the survival and development of discrete neuronal subpopulations, and the receptor tyrosine kinases TIE1 (MIM 600222) and TIE2 (MIM 600221) play a critical role in the development of normal blood vessels.[supplied by OMIM

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen MUSK
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description muscle, skeletal, receptor tyrosine kinase
Formulation In 1X PBS, pH 7.4
Gene MUSK
Gene Accession No. MUSK
Gene Alias MGC126323, MGC126324
Gene Symbols MUSK
Host Species Rabbit
Immunogen A synthetic peptide of human MUSK
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4593
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.