Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mevalonate (diphospho) decarboxylase, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017872 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-872 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-872 Supplier Abnova Corporation Supplier No. H00004597D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human MVD protein.

The enzyme mevalonate pyrophosphate decarboxylase catalyzes the conversion of mevalonate pyrophosphate into isopentenyl pyrophosphate in one of the early steps in cholesterol biosynthesis. It decarboxylates and dehydrates its substrate while hydrolyzing ATP. [provided by RefSeq

Sequence: MASEKPLAAVTCTAPVNIAVIKYWGKRDEELVLPINSSLSVTLHQDQLKTTTTAVISKDFTEDRIWLNGREEDVGQPRLQACLREIRCLARKRRNSRDGDPLPSSLSCKVHVASVNNFPTAAGLASSAAGYACLAYTLARVYGVESDLSEVARRGSGSACRSLYGGFVEWQMGEQADGKDSIARQVAPESHWPELRVLILVVSAEKKLTGSTVGMRASVETSPLLRFRAESVVPARMAEMARCIRERDFPSFAQLTMKDSNQFHATCLDTFPPISYLNAISWRIIHLVHRFNAHHGDTKVAYTFDAGPNAVIFTLDDTVAEFVAAVWHGFPPGSNGDTFLKGLQVRPAPLSAELQAALAMEPTPGGVKYIIVTQVGPGPQILDDPCAHLLGPDGLPKPAA

Specifications

Antigen mevalonate (diphospho) decarboxylase
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human MVD protein.
Formulation PBS with no preservative; pH 7.4
Gene MVD
Gene Accession No. NM_002461.1
Gene Alias FP17780/MPD
Gene Symbols MVD
Host Species Rabbit
Immunogen MVD (NP_002452.1, 1 a.a. ∼ 400 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4597
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.