Learn More
myxovirus (influenza virus) resistance 1, interferon-inducible protein p78 (mouse), Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
In mouse, the interferon-inducible Mx protein is responsible for a specific antiviral state against influenza virus infection. The protein encoded by this gene is similar to the mouse protein as determined by its antigenic relatedness, induction conditions, physicochemical properties, and amino acid analysis. This cytoplasmic protein is a member of both the dynamin family and the family of large GTPases. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq
Specifications
Specifications
| Antigen | myxovirus (influenza virus) resistance 1, interferon-inducible protein p78 (mouse) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a full-length human MX1 protein. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | MX1 |
| Gene Accession No. | NM_002462 |
| Gene Alias | IFI-78K/IFI78/MX/MxA |
| Gene Symbols | MX1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.