Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-MxB, Polyclonal
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ ABF1059
Description
Specifically detects MxB Clone: in Human samples, and it is validated for Western Blotting
Interferon-induced GTP-binding protein Mx2 (UniProt P20592; also known as Interferon-regulated resistance GTP-binding protein MxB, Myxovirus resistance protein 2, p78-related protein, Second interferon-induced protein p78) is encoded by the MX2 (also known as MXB) gene (Gene ID 4600) in human. The interferon-inducible myxovirus resistance (Mx) proteins belong to the family of large GTPases and are highly homologous with dynamins within their GTP-binding domain. MxA/Mx1 is cytosolic, while two MxB/Mx forms exist, a 78kDa nuclear form and a 76kDa cytosolic form lacking the N-terminal nuclear localization signal (NLS). Human MxA protein mediates resistance to influenza and several other viruses, while human MxB is also reported to inhibit HIV-1 infection by reducing the level of integrated viral DNA. In addition, HIV-1 with Ala88 mutation in the capsid protein (CA) is found to escape MxB restriction, as a result of impaired CA interaction with the host peptidylprolyl isomerase cyclophilin A (CypA). Likewise, CypA deletion results in the loss of MxB inhibition of HIV-1.
Specifications
MxB | |
Polyclonal | |
Rabbit polyclonal serum with 0.05% sodium azide. | |
MX2 | |
GST-tagged recombinant protein corresponding to the Dynamin-type G domain of human MxB. | |
100 μL | |
Inflammation, Immunology | |
NP_002454 | |
Human | |
Serum |
Western Blot | |
Unconjugated | |
P20592 | |
Rabbit | |
Unpurified | |
RUO | |
Primary | |
Expected to react with both spliced isoforms. | |
−20°C for one year |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction