Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myb Mouse anti-Human, Mouse, Clone: 1-1, MilliporeSigma™

Catalog No. 05175MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-175-MI 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-175-MI Supplier MilliporeSigma Supplier No. 05175
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Myb Antibody, clone 1-1 is a Mouse Monoclonal Antibody for detection of Myb also known as Avian myeloblastosis viral (v-myb) oncogene homolog 2 c-myb protein (140 AA) and has been validated in immunohistochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myb
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1-1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-glycine, pH 7.4, 0.15MNaCl and 0.05% sodium azide. Frozen solution.
Gene Accession No. NM_005375.2
Gene Symbols MYB, C-myb, c-myb13A_CDS, c-myb8B_CDS, c-myb_CDS, v-myb, c-myb14A_CDS, Cmyb, c-myb, efg, c-myb10A_CDS
Host Species Mouse
Immunogen Recombinant murine Myb purified by SDS-PAGE (Nicot, C., 2001) Clone 1-1.
Purification Method Ammonium sulfate precipitation, gel filtration, and ion-exchange chromatography.
Quantity 250 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.