Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myc Tag Mouse anti-All, AlexaFluor™488, Clone: 9E10, MilliporeSigma™

Catalog No. 16308MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-308-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-308-MI Supplier MilliporeSigma Supplier No. 16308
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Myc Tag Antibody, clone 9E10, Alexa Fluor 488 conjugate is validated for use in western blot, IC for the detection of Myc Tag.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myc Tag
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 9E10
Conjugate Alexa Fluor 488
Gene Accession No. NM_002467.3
Gene Symbols MYC, C-MYC, c-Myc
Host Species Mouse
Immunogen Synthetic peptide corresponding to amino acids 408-437 of the Leucine zipper region of human Myc (AEEQKLISEEDLLRKRREQLKHKLEQ-LRNS)
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species All species
Content And Storage 4°C for one year
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.