Learn More
Mycobacterium tuberculosis, FITC, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA128997
Description
Tuberculosis is a chronic bacterial infection which remains one of the leading killers of young adults world wide. Mycobacterium tuberculosis enters the alveoli by airborne transmission. They are engulfed by alveolar macrophages but resist destruction. Although the exact mechanism is not known, it is suspected that that Mycrobacterium tuberculosis survives by preventing the fusion of the phagosome and lysosome. As the bacterium accumulates it forms a tubercle, spreads to the lymph nodes and becomes a systemic infection. Although treatments for tuberculosis do exist, new strains of Mycobacterium tuberculosis that appear to have multiple-drug resistance (MDR) pose a considerable threat to public health.
PA1-28997 detects Mycobacterium tuberculosis from bacteria samples.PA1-28997 has been successfully used in ELISA and immunohistochemistry (frozen tissue) applications.The PA1-28997 immunogen is: Purified PPD.
Specifications
Mycobacterium tuberculosis | |
Polyclonal | |
FITC | |
Rabbit | |
IgG fraction | |
RUO | |
Bacteria | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
4mg/mL | |
M. tuberculosis, myobacter | |
Purified PPD. | |
500 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.