Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mycoplasma bovis Mouse anti-Bovine, Clone: MYB163, MilliporeSigma™

Catalog No. MAB970MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB970MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB970MI Supplier MilliporeSigma Supplier No. MAB970
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Mycoplasma bovis Antibody, Strain M23, clone MYB163 detects level of Mycoplasma bovis and has been published and validated for use in ELISA, western blot, immunohistochemistry, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mycoplasma bovis
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone MYB163
Conjugate Unconjugated
Formulation Ascites fluid contain 10% buffered formalin.
Host Species Mouse
Immunogen Mycoplasma bovis strain M23.
Quantity 100 μL
Primary or Secondary Primary
Target Species Bovine
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.