Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MyD88 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB16527MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB16527MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB16527MI Supplier MilliporeSigma Supplier No. AB16527
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-MyD88 Antibody, C-terminus is an antibody against MyD88 for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MyD88
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in PBS containing 0.02% sodium azide.
Gene Symbols MYD88
Host Species Rabbit
Immunogen Rabbit anti-MyD88 (C-terminus) polyclonal antibody was raised against a peptide corresponding to amino acids 279 to 296 of human MyD88 (Bonnert et al. 1997), which are identical to those of mouse.
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_002468.3
Target Species Human, Mouse
Content And Storage 2-8°C in undiluted aliquots for up to 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.