Learn More
MilliporeSigma™ anti-Myelin Basic Protein, aa 82-87 (MBP), ™ (Chemicon™)
Supplier: MilliporeSigma™ MAB386/DEL
Description
ELISA, Immunocytochemistry, Immunohistochemistry, Radioimmunoassay, Western Blotting
Specifications
Myelin Basic Protein, aa 82-87 (MBP) | |
Unconjugated | |
Bovine myelin basic protein. | |
RUO | |
Primary | |
Ascites |
Monoclonal | |
Rat | |
1 mL | |
Neuroscience | |
Bovine, Chicken, Guinea Pig, Human, Mouse, Rabbit, Rat, Sheep | |
IgG2a |