Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myelin Basic Protein Rat anti-Bovine, Chicken, Guinea Pig, Human, Mouse, Rat, Sheep, Rabbit, Clone: 12, MilliporeSigma™

Catalog No. MAB386MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB386MI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB386MI Supplier MilliporeSigma Supplier No. MAB386
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-Myelin Basic Protein Antibody, a.a. 82-87 is an antibody against Myelin Basic Protein for use in ELISA, immunocytochemistry, immunohistochemistry, RIA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myelin Basic Protein
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone 12
Conjugate Unconjugated
Formulation Liquid containing 0.09% sodium azide.
Gene Accession No. NM_001025094.1
Gene Symbols MBP, MGC99675, Golli-mbp
Host Species Rat
Immunogen Bovine myelin basic protein
Purification Method Unpurified
Quantity 1 mL
Primary or Secondary Primary
Target Species Bovine, Chicken, Guinea Pig, Human, Mouse, Rabbit, Rat, Sheep
Content And Storage −20°C for up to 12 months.
Form Ascites
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.