Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myelin Basic Protein Rat anti-Mammals, Clone: 14, MilliporeSigma™

Catalog No. MAB3951MLMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3951MLMI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3951MLMI Supplier MilliporeSigma Supplier No. MAB3951ML
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Detect Myelin Basic Protein using this Anti-Myelin Basic Protein Antibody, a.a. 36-50, clone 14 validated for use in ELISA, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myelin Basic Protein
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 14
Conjugate Unconjugated
Formulation Tissue culture supernatant. Liquid containing 0.09% sodium azide.
Gene Accession No. NM_001025094.1; NM_001025098.1; NM_001025081.1; NM_001025090.1; NM_001025092.1; NM_001025100.1; NM_001025101.1; NM_002385.2
Gene Symbols MBP, MGC99675, Golli-mbp
Host Species Rat
Immunogen Bovine myelin basic protein.
Quantity 1 mL
Primary or Secondary Primary
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Supernatant
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.