Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myelin Basic Protein Mouse anti-Bovine, Guinea Pig, Human, Rat, Rabbit, Sheep, Clone: 2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB381
Description
Anti-Myelin Basic Protein Antibody, a.a. 119-131, clone 2 is an antibody against Myelin Basic Protein for use in ELISA, immunohistochemistry.
Specifications
Myelin Basic Protein | |
Monoclonal | |
Unconjugated | |
NM_001025094.1; NM_001025098.1; NM_001025081.1; NM_001025090.1; NM_001025092.1; NM_001025100.1; NM_001025101.1; NM_002385.2 | |
Mouse | |
2 mL | |
Bovine, Guinea Pig, Human, Rabbit, Rat, Sheep | |
Supernatant |
ELISA, Immunohistochemistry | |
2 | |
Tissue culture supernatant. Liquid in 0.2M Tris/HCl, pH 7.4 with 5-10% fetal calf serum and 0.09% sodium azide. | |
MBP, MGC99675, Golli-mbp | |
Bovine myelin basic protein. | |
Primary | |
−20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction